COM448614230 Combi-Blocks, Inc.

3-(Ethyl(methyl)carbamoyl)phenylboronic acid

  • MDL number MFCD08056369
  • Cas Number : 871333-07-2
  • Molecular Weight : 207.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3098
Availability details for
Quantity
  • Availability
  • Add to Cart