COM448614233 Combi-Blocks, Inc.

4-(3-Chloropropylcarbamoyl)phenylboronic acid

  • MDL number MFCD08235077
  • Cas Number : 874460-03-4
  • Molecular Weight : 241.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3102
Availability details for
Quantity
  • Availability
  • Add to Cart