COM448614241 Combi-Blocks, Inc.

N-(3-Bromopropyl) 3-boronobenzamide

  • MDL number MFCD06659892
  • Cas Number : 850567-42-9
  • Molecular Weight : 285.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3150
Availability details for
Quantity
  • Availability
  • Add to Cart