COM448614249 Combi-Blocks, Inc.

4-(Phenethylcarbamoyl)phenylboronic acid

  • MDL number MFCD09027209
  • Cas Number : 330793-46-9
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3201
Availability details for
Quantity
  • Availability
  • Add to Cart