COM448614449 Combi-Blocks, Inc.

4-Boronobenzenesulfonic acid

  • MDL number MFCD09027231
  • Cas Number : 913836-00-7
  • Molecular Weight : 202.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3544
Availability details for
Quantity
  • Availability
  • Add to Cart