COM448614560 Combi-Blocks, Inc.

2-Methylpyridine-3-boronic acid

  • MDL number MFCD07368825
  • Cas Number : 899436-71-6
  • Molecular Weight : 136.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3922

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3922
Availability details for
Quantity
  • Availability
  • Add to Cart