COM448614781 Combi-Blocks, Inc.

3-Amino-5-(pyrrolidinocarbonyl)phenylboronic acid

  • MDL number MFCD22192447
  • Cas Number : 2096336-11-5
  • Molecular Weight : 234.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4431
Availability details for
Quantity
  • Availability
  • Add to Cart