COM448614850 Combi-Blocks, Inc.

[3-(1-{[(tert-Butoxy)carbonyl]amino}-2-methylpropan-2-yl)phenyl]boronic acid

  • MDL number MFCD28096631
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4641

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4641
Availability details for
Quantity
  • Availability
  • Add to Cart