COM448614878 Combi-Blocks, Inc.

4-(3-Methylphenoxy)phenylboronic acid

  • MDL number MFCD28024838
  • Cas Number : 1029438-39-8
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4718

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4718
Availability details for
Quantity
  • Availability
  • Add to Cart