COM448614918 Combi-Blocks, Inc.

3-(4-BOC-Piperazino)-5-chlorophenylboronic acid

  • MDL number MFCD28369561
  • Cas Number : 2377609-41-9
  • Molecular Weight : 340.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4866
Availability details for
Quantity
  • Availability
  • Add to Cart