COM448614919 Combi-Blocks, Inc.

4-(4-BOC-Piperazino)-3-chlorophenylboronic acid

  • MDL number MFCD28122668
  • Molecular Weight : 340.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4868

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4868
Availability details for
Quantity
  • Availability
  • Add to Cart