COM448614921 Combi-Blocks, Inc.

5-(4-BOC-Piperazinomethyl)thiophene-2-boronic acid

  • MDL number MFCD20527083
  • Cas Number : 2377609-49-7
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4877
Availability details for
Quantity
  • Availability
  • Add to Cart