COM448614923 Combi-Blocks, Inc.

4-{2-[(tert-Butyldimethylsilyl)oxy]ethoxy}phenylboronic acid

  • MDL number MFCD28101632
  • Cas Number : 225385-14-8
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4892
Availability details for
Quantity
  • Availability
  • Add to Cart