COM448614929 Combi-Blocks, Inc.

Bis(4-methoxynaphthalen-1-yl)borinic acid

  • MDL number MFCD28101634
  • Molecular Weight : 342.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4906
Availability details for
Quantity
  • Availability
  • Add to Cart