COM448614931 Combi-Blocks, Inc.

3-(Benzyloxy)-2-chlorophenylboronic acid

  • MDL number MFCD28122669
  • Cas Number : 2377611-50-0
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4910
Availability details for
Quantity
  • Availability
  • Add to Cart