COM448614933 Combi-Blocks, Inc.

4-Fluoro-5-formyl-2-methylphenylboronic acid

  • MDL number MFCD28166375
  • Cas Number : 2377606-69-2
  • Molecular Weight : 182.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4925
Availability details for
Quantity
  • Availability
  • Add to Cart