COM448614936 Combi-Blocks, Inc.

2-(2-Chloro-6-methoxyphenyl)-6-methoxyphenylboronic acid

  • MDL number MFCD28127179
  • Molecular Weight : 292.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4934

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4934
Availability details for
Quantity
  • Availability
  • Add to Cart