COM448614937 Combi-Blocks, Inc.

3-Bromo-5-chloro-2-fluorophenylboronic acid

  • MDL number MFCD28143924
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4954

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4954
Availability details for
Quantity
  • Availability
  • Add to Cart