COM448615023 Combi-Blocks, Inc.

1-p-Tolylpyrazole-4-boronic acid

  • MDL number MFCD09972102
  • Cas Number : 1072945-92-6
  • Molecular Weight : 202.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5121

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5121
Availability details for
Quantity
  • Availability
  • Add to Cart