COM448615078 Combi-Blocks, Inc.

4-(1-BOC-piperidin-4-yloxy)-2-methoxyphenylboronic acid

  • MDL number MFCD10699635
  • Cas Number : 1072946-29-2
  • Molecular Weight : 351.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5214
Availability details for
Quantity
  • Availability
  • Add to Cart