COM448615210 Combi-Blocks, Inc.

2,3-Difluoro-5-methoxyphenylboronic acid

  • MDL number MFCD27935900
  • Cas Number : 2230876-12-5
  • Molecular Weight : 187.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5531
Availability details for
Quantity
  • Availability
  • Add to Cart