COM448615227 Combi-Blocks, Inc.

[6-Chloro-5-(trifluoromethyl)pyridin-3-yl]boronic acid

  • MDL number MFCD18254528
  • Cas Number : 2096336-32-0
  • Molecular Weight : 225.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5573

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5573
Availability details for
Quantity
  • Availability
  • Add to Cart