COM448615239 Combi-Blocks, Inc.

1-(2-Hydroxyethyl)pyrazole-4-boronic acid

  • MDL number MFCD16621237
  • Cas Number : 1086063-70-8
  • Molecular Weight : 155.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5621

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5621
Availability details for
Quantity
  • Availability
  • Add to Cart