COM448615263 Combi-Blocks, Inc.

2,3-Difluoro-5-(methoxycarbonyl)phenylboronic acid

  • MDL number MFCD28127180
  • Molecular Weight : 215.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5682

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5682
Availability details for
Quantity
  • Availability
  • Add to Cart