COM448615267 Combi-Blocks, Inc.

[6-Methoxy-5-(trifluoromethyl)pyridin-3-yl]boronic acid

  • MDL number MFCD18254530
  • Cas Number : 1420830-61-0
  • Molecular Weight : 220.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5712

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5712
Availability details for
Quantity
  • Availability
  • Add to Cart