COM448615725 Combi-Blocks, Inc.

2-Fluoro-6-(hydroxymethyl)phenylboronic acid

  • MDL number MFCD18383507
  • Cas Number : 1246633-54-4
  • Molecular Weight : 169.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-6986
Availability details for
Quantity
  • Availability
  • Add to Cart