COM448616151 Combi-Blocks, Inc.

2-Picoline-5-boronic acid hydrate

  • MDL number MFCD06657901
  • Cas Number : 1072952-30-7
  • Molecular Weight : 154.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8495
Availability details for
Quantity
  • Availability
  • Add to Cart