COM448616228 Combi-Blocks, Inc.

Vinylboronic acid

  • MDL number MFCD02258951
  • Cas Number : 4363-34-2
  • Molecular Weight : 71.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8602
Availability details for
Quantity
  • Availability
  • Add to Cart