COM448616264 Combi-Blocks, Inc.

1-[1,3]-Dioxolan-2-ylmethyl-1h-pyrazole-4-boronic acid

  • MDL number MFCD11042451
  • Cas Number : 947533-27-9
  • Molecular Weight : 198.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8731
Availability details for
Quantity
  • Availability
  • Add to Cart