COM448616266 Combi-Blocks, Inc.

4-N-Boc-amino-3-methoxy-phenylboronic acid

  • MDL number MFCD03426361
  • Cas Number : 669713-95-5
  • Molecular Weight : 267.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8743

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8743
Availability details for
Quantity
  • Availability
  • Add to Cart