COM448616276 Combi-Blocks, Inc.

1-(Tetrahydropyran-2-yl)pyrazole-5-boronic acid

  • MDL number MFCD11046121
  • Cas Number : 1105511-68-9
  • Molecular Weight : 196.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8781
Availability details for
Quantity
  • Availability
  • Add to Cart