COM448616279 Combi-Blocks, Inc.

(5-Aminopyridin-3-yl)boronic acid

  • MDL number MFCD07374883
  • Cas Number : 1169748-84-8
  • Molecular Weight : 137.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8793

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8793
Availability details for
Quantity
  • Availability
  • Add to Cart