COM448616388 Combi-Blocks, Inc.

4-(2-Hydroxyethoxy)phenylboronic acid

  • MDL number MFCD22414430
  • Cas Number : 221006-65-1
  • Molecular Weight : 182.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1016

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1016
Availability details for
Quantity
  • Availability
  • Add to Cart