COM448616402 Combi-Blocks, Inc.

2-(Isobutylsulfanyl)phenylboronic acid

  • MDL number MFCD28385194
  • Cas Number : 2377606-61-4
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1061
Availability details for
Quantity
  • Availability
  • Add to Cart