COM448616798 Combi-Blocks, Inc.

4-(Morpholine-4-carbonyl)bromobenzene

  • MDL number MFCD00447938
  • Cas Number : 127580-92-1
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4410
Availability details for
Quantity
  • Availability
  • Add to Cart