COM448616960 Combi-Blocks, Inc.

Methyl 5-bromo-2-nitrobenzoate

  • MDL number MFCD00092886
  • Cas Number : 883554-93-6
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4599

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4599
Availability details for
Quantity
  • Availability
  • Add to Cart