COM448617092 Combi-Blocks, Inc.

5-(3-bromophenyl)pentanoic acid

  • MDL number MFCD18459232
  • Cas Number : 857480-35-4
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4761

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4761
Availability details for
Quantity
  • Availability
  • Add to Cart