COM448617122 Combi-Blocks, Inc.

3-chloro-5-methoxybenzonitrile

  • MDL number MFCD11505060
  • Cas Number : 473923-96-5
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4797
Availability details for
Quantity
  • Availability
  • Add to Cart