COM448617198 Combi-Blocks, Inc.

3-(1H-Indol-5-yl)benzoic acid

  • MDL number MFCD03839964
  • Cas Number : 886363-16-2
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-4880
Availability details for
Quantity
  • Availability
  • Add to Cart