COM448617399 Combi-Blocks, Inc.

3-Methoxy-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD08458145
  • Cas Number : 868167-61-7
  • Molecular Weight : 201.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5107
Availability details for
Quantity
  • Availability
  • Add to Cart