COM448617529 Combi-Blocks, Inc.

4-Bromo-N-cyclopropylnaphthalene-1-carboxamide

  • MDL number MFCD21609553
  • Cas Number : 1365271-49-3
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5293

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5293
Availability details for
Quantity
  • Availability
  • Add to Cart