COM448617533 Combi-Blocks, Inc.

3-Methoxy-4-propoxybenzoic acid

  • MDL number MFCD03658025
  • Cas Number : 3535-32-8
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5297

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5297
Availability details for
Quantity
  • Availability
  • Add to Cart