COM448617545 Combi-Blocks, Inc.

methyl 4-(cyclopropylcarbamoyl)benzoate

  • MDL number MFCD17579950
  • Cas Number : 880166-77-8
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5314

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5314
Availability details for
Quantity
  • Availability
  • Add to Cart