COM448617547 Combi-Blocks, Inc.

Methyl 4-(propylcarbamoyl)benzoate

  • MDL number MFCD09418578
  • Cas Number : 216162-45-7
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5316

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5316
Availability details for
Quantity
  • Availability
  • Add to Cart