COM448617561 Combi-Blocks, Inc.

1-(4-Bromo-3-methoxyphenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD22192463
  • Cas Number : 1373232-79-1
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5335
Availability details for
Quantity
  • Availability
  • Add to Cart