COM448617570 Combi-Blocks, Inc.

4-Bromo-3-chlorobenzamide

  • MDL number MFCD17779957
  • Cas Number : 1228826-41-2
  • Molecular Weight : 234.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5349
Availability details for
Quantity
  • Availability
  • Add to Cart