COM448617575 Combi-Blocks, Inc.

4-(Ethylcarbamoyl)benzoic acid

  • MDL number MFCD12173485
  • Cas Number : 167627-38-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5359

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5359
Availability details for
Quantity
  • Availability
  • Add to Cart