COM448617576 Combi-Blocks, Inc.

4-(Diethylcarbamoyl)benzoic acid

  • MDL number MFCD12173474
  • Cas Number : 71888-24-9
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5361
Availability details for
Quantity
  • Availability
  • Add to Cart