COM448617591 Combi-Blocks, Inc.

2-tert-Butyl-3-hydroxy-5-methoxy-3H-isoindol-1-one

  • MDL number MFCD22375029
  • Cas Number : 1381944-83-7
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5387
Availability details for
Quantity
  • Availability
  • Add to Cart