COM448617593 Combi-Blocks, Inc.

4-(Piperidinocarbonyl)benzonitrile

  • MDL number MFCD06867205
  • Cas Number : 160094-26-8
  • Molecular Weight : 214.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5389
Availability details for
Quantity
  • Availability
  • Add to Cart